Assay Details
| Gene Symbol: Myoz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021508.3 | NP_067483.1 | 4 - 5 | 621 | 150 | |
| GenBank mRNA | AJ005620.1 | - | 4 - 5 | 561 | 150 | |
| AK009077.1 | - | 4 - 5 | 585 | 150 | ||
| AK041896.1 | - | 4 - 5 | 577 | 150 | ||
| AY013298.1 | - | 4 - 5 | 539 | 150 | ||
| BC024660.1 | - | 4 - 5 | 677 | 3967841 | 150 | |
More Information
Updated Mapping Information
Set Membership: |
|

Myoz1
Genomic Map