Assay Details
| Gene Symbol: Usp54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030180.2 | NP_084456.2 | 13 - 14 | 2620 | 60 | |
| GenBank mRNA | CF535703.1 | - | 2 - 3 | 447 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Usp54
Genomic Map