Assay Details
| Gene Symbol: Usp54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030180.2 | NP_084456.2 | 17 - 18 | 3242 | 91 | |
| GenBank mRNA | CK638913.1 | - | 4 - 5 | 595 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Usp54
Genomic Map