Assay Details
| Gene Symbol: H2-Ob | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK155144.1 | - | 2 - 3 | 380 | 83 | |
| AK155650.1 | - | 2 - 3 | 430 | 83 | ||
| AK172191.1 | - | 2 - 3 | 435 | 83 | ||
| BC100709.1 | - | 2 - 3 | 356 | 83 | ||
| KJ650230.1 | - | 2 - 3 | 363 | 83 | ||
More Information
Set Membership: |
|

H2-Ob
Genomic Map