Assay Details
| Gene Symbol: Sbf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177324.2 | NP_796298.2 | 20 - 21 | 2685 | 76 | |
| GenBank mRNA | AK046622.1 | - | 20 - 21 | 2673 | 76 | |
| CF745183.1 | - | 1 - 2 | 163 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sbf2
Genomic Map