Assay Details
| Gene Symbol: Knstrn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026412.3 | NP_080688.2 | 4 - 5 | 575 | 70 | |
| GenBank mRNA | AK006328.1 | - | 4 - 5 | 576 | 70 | |
| AK013184.1 | - | 4 - 5 | 323 | 70 | ||
| AK028590.1 | - | 4 - 5 | 376 | 70 | ||
| AK036374.1 | - | 4 - 5 | 364 | 70 | ||
| BC031709.1 | - | 3 - 4 | 283 | 4021550 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Knstrn
Genomic Map