Assay Details
| Gene Symbol: Knstrn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026412.3 | NP_080688.2 | 6 - 7 | 778 | 100 | |
| GenBank mRNA | AK012148.1 | - | 6 - 7 | 578 | 100 | |
| AK028590.1 | - | 6 - 7 | 579 | 100 | ||
| AK036374.1 | - | 6 - 7 | 567 | 100 | ||
| BC031709.1 | - | 5 - 6 | 486 | 4021550 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Knstrn
Genomic Map