Assay Details
| Gene Symbol: Rras2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025846.2 | NP_080122.2 | 3 - 4 | 573 | 89 | |
| GenBank mRNA | AK078870.1 | - | 3 - 4 | 574 | 89 | |
| AK172037.1 | - | 3 - 4 | 565 | 89 | ||
| AK172113.1 | - | 3 - 4 | 559 | 89 | ||
| AK216768.1 | - | 1 - 2 | 86 | 89 | ||
| BC003871.1 | - | 3 - 4 | 524 | 3492159 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rras2
Genomic Map