Assay Details
| Gene Symbol: Itpripl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163528.1 | NP_001157000.1 | 1 - 2 | 64 | 89 | |
| GenBank mRNA | AK006667.1 | - | 1 - 2 | 65 | 89 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001163528.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P676255 | AK028913.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P676255 | BC130218.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P676255 | NM_001163528.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P676255 | AK006667.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P676255 | NM_001163527.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P676255 | AK028913.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P676255 | BC130218.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P676255 | NM_001163528.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P676255 | AK006667.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P676255 | NM_001163527.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 240969 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 294695 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 542085 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P178361 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P676255 | AK028913.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P676255 | BC130218.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P676255 | NM_001163528.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P676255 | AK006667.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P676255 | NM_001163527.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P676255 | AK028913.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P676255 | BC130218.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P676255 | NM_001163528.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P676255 | AK006667.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P676255 | NM_001163527.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P263407 | AK028913.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P263407 | NM_001163528.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P263407 | NM_001163527.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P676255 | AK028913.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P676255 | BC130218.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P676255 | NM_001163528.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P676255 | AK006667.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P676255 | NM_001163527.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P676255 | AK028913.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P676255 | BC130218.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P676255 | NM_001163528.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P676255 | AK006667.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P676255 | NM_001163527.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P109916 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21611840 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21611834 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01357460 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21611848 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21611836 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21611838 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21611841 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21611849 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21611843 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10487365 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10487362 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10487363 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10487361 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10487364 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1431234_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10487359 | - | - |
| Mouse Genome 430 2.0 Array | 1431234_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1431234_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 73338_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 73338_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200038958.mm.1 | NM_001163528.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200038958.mm.1 | NM_001163528.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200038958.mm.1 | TC0200004548.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200038953.mm.1 | NM_001163527.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200038953.mm.1 | NM_001163527.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200038953.mm.1 | NM_001163528.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200038953.mm.1 | NM_001163528.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200038953.mm.1 | TC0200004548.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200038955.mm.1 | NM_001163527.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200038955.mm.1 | NM_001163527.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200038955.mm.1 | NM_001163528.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200038955.mm.1 | NM_001163528.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200038955.mm.1 | TC0200004548.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200038954.mm.1 | NM_001163527.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200038954.mm.1 | NM_001163527.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200038954.mm.1 | NM_001163528.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200038954.mm.1 | NM_001163528.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200038954.mm.1 | TC0200004548.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200038956.mm.1 | NM_001163527.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200038956.mm.1 | NM_001163527.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200038956.mm.1 | NM_001163528.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200038956.mm.1 | NM_001163528.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200038956.mm.1 | TC0200004548.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1431234_at | - | - |
| Mouse Expression Set 430 | 1431234_PM_at | - | - |
| Murine Genome U74v2 Set | 131609_at | - | - |
| Murine Genome U74v2 Set | 131609_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431234_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431234_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 729003 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2737831 | - | - |

Itpripl1
Genomic Map