Assay Details
| Gene Symbol: Prrg4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178695.5 | NP_848810.3 | 3 - 4 | 599 | 116 | |
| GenBank mRNA | AK035530.1 | - | 3 - 4 | 424 | 116 | |
| AK037087.1 | - | 3 - 4 | 519 | 116 | ||
| AK038703.1 | - | 3 - 4 | 599 | 116 | ||
| AK166405.1 | - | 2 - 3 | 560 | 116 | ||
| BC099390.1 | - | 3 - 4 | 516 | 30715383 | 116 | |
| BC115470.2 | - | 3 - 4 | 345 | 40100436 | 116 | |
| BC151011.1 | - | 3 - 4 | 529 | 9087924 | 116 | |
| BC151019.1 | - | 3 - 4 | 529 | 9087932 | 116 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prrg4
Genomic Map