Assay Details
| Gene Symbol: Ppapdc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080963.1 | NP_001074432.1 | 2 - 3 | 185 | 65 | |
| GenBank mRNA | AK081292.1 | - | 2 - 3 | 249 | 65 | |
| BC120497.1 | - | 2 - 3 | 185 | 40129420 | 65 | |
| BC144735.1 | - | 2 - 3 | 185 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ppapdc1a
Genomic Map