| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700037346.mm.1 |
NM_001143802.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700037346.mm.1 |
NM_001143802.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700037346.mm.1 |
TC0700004444.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700037349.mm.1 |
NM_001143802.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700037349.mm.1 |
NM_001143802.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700037349.mm.1 |
TC0700004444.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700037345.mm.1 |
NM_001143802.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700037345.mm.1 |
NM_001143802.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700037345.mm.1 |
TC0700004444.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700037347.mm.1 |
NM_001143802.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700037347.mm.1 |
NM_001143802.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700037347.mm.1 |
TC0700004444.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700037344.mm.1 |
NM_001143802.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700037344.mm.1 |
NM_001143802.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700037344.mm.1 |
TC0700004444.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700037348.mm.1 |
NM_001143802.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700037348.mm.1 |
NM_001143802.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700037348.mm.1 |
TC0700004444.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700037343.mm.1 |
TC0700004444.mm.1 |
MTA TC |