| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.0 ST Array |
10568706 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10568708 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10568707 |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1446062_PM_at |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1443760_PM_at |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1457806_PM_at |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10568705 |
- |
- |
| Mouse Genome 430 2.0 Array |
1443760_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1446062_PM_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1457806_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1457806_PM_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1443760_PM_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1446062_at |
- |
- |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array |
627214_at |
- |
- |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array |
627214_PM_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR0700037341.mm.1 |
NM_001143802.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700037341.mm.1 |
NM_001143802.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700037341.mm.1 |
TC0700004444.mm.1 |
MTA TC |
| Mouse Expression Set 430 |
1457806_PM_at |
- |
- |
| Mouse Expression Set 430 |
1457806_at |
- |
- |
| Mouse Expression Set 430 |
1446062_at |
- |
- |
| Mouse Expression Set 430 |
1443760_PM_at |
- |
- |
| Mouse Expression Set 430 |
1443760_at |
- |
- |
| Mouse Expression Set 430 |
1446062_PM_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1443760_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1457806_PM_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1446062_PM_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1457806_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1446062_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1443760_PM_at |
- |
- |