Assay Details
| Gene Symbol: Folr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252552.1 | NP_001239481.1 | 2 - 3 | 333 | 93 | |
| GenBank mRNA | BC138795.1 | - | 1 - 2 | 50 | 8861817 | 93 |
| BC145414.1 | - | 1 - 2 | 50 | 9053972 | 93 | |
| CJ079096.1 | - | 2 - 3 | 268 | 93 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC138795.1, BC145414.1, NM_001252552.1 transcripts.
Set Membership: |
|

Folr1
Genomic Map