Assay Details
| Gene Symbol: Olfr676 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_147095.1 | NP_667306.1 | 1 | 652 | 120 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2005621 | NM_147095.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2005620 | NM_147095.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P102321 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2005620 | NM_147095.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2005621 | NM_147095.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P389317 | NM_147095.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P102321 | - | - |
| Whole Mouse Genome G4122A | A_51_P389317 | NM_147095.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P102321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390457 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390476 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390409 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06341228 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06516263 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390473 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06345692 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06089438 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390453 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390460 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390469 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390452 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06626961 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390427 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390455 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390430 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390472 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390423 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390417 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390422 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390464 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390439 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390450 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390432 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390414 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390412 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390415 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390413 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390425 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06089451 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390426 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390457 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390447 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390467 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390446 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390428 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390463 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390449 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390438 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390437 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390474 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390466 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390440 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390443 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10555904 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10555903 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 524865 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2360082 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2360082 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002360082 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1256438 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1256438 | - | - |
| mouse-6 v1.1 (Illumina) | scl9382.1.1_106-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2360082 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl9382.1.1_106-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2360082 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1256438 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1256438 | - | - |

Olfr676
Genomic Map