Assay Details
| Gene Symbol: Olfr1164 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146641.2 | NP_666852.2 | 1 | 150 | 123 | |
| GenBank mRNA | BC132589.1 | - | 1 | 150 | 40130866 | 123 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2132741 | NM_146641.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2132741 | BC132589.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 742260 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 256299 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 212151 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 161805 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 622289 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 150946 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 154711 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 1720 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 66353 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 623 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P133322 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P141988 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2132741 | NM_146641.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2132741 | BC132589.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P490618 | NM_146641.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P490618 | BC132589.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21408521 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21408517 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21408518 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21408520 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21408519 | - | - |
| Whole Mouse Genome G4122A | A_51_P490618 | NM_146641.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P490618 | BC132589.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408473 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408516 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408485 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408519 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408480 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408492 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408493 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408487 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408456 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408451 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408461 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01148405 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408482 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408455 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408490 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408521 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408476 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408518 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408511 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408481 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02237638 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408498 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02237627 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408464 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408459 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408488 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408520 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408505 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02237661 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01652900 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408507 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408494 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408473 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408510 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408472 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408475 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408465 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408483 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408454 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408462 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408452 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408502 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408515 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408496 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408469 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408479 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408513 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408466 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10484717 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10484716 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200033921.mm.1 | NM_146641.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200033921.mm.1 | NM_146641.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200033921.mm.1 | TC0200004007.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 817098 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3290113 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3290113 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003290113 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1225305 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1225305 | - | - |
| mouse-6 v1.1 (Illumina) | scl4303.1.1_75-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3290113 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl4303.1.1_75-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3290113 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1225305 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1225305 | - | - |

Olfr1164
Genomic Map