Assay Details
Gene Symbol: Olfr655 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146820.2 | NP_667031.2 | 1 | 822 | 68 | |
GenBank mRNA | BC114416.1 | - | 1 | 821 | 40040785 | 68 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2041559 | NM_146820.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2041559 | BC114416.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P126547 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2041559 | NM_146820.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2041559 | BC114416.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P140263 | NM_146820.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25388738 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25388743 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25388774 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25388739 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25388775 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25388765 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25388781 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25388767 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25388789 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25388749 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25388746 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25388787 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25388772 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25388769 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25388741 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25388740 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25388754 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25388753 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25388751 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25388773 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25388784 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25388755 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25388770 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25388761 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25388785 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25388759 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25388763 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25388748 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25388780 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25388747 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25388750 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25388783 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25388757 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25388764 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25388777 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P126547 | - | - |
Whole Mouse Genome G4122A | A_51_P140263 | NM_146820.2 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P117560 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P126547 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06519857 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06081049 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06081064 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06519854 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25388682 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25388751 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25388691 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25388677 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25388775 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25388741 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25388763 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25388785 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25388743 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25388711 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25388703 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25388694 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25388681 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25388697 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25388720 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25388797 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25388701 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25388725 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25388706 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25388712 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25388787 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25388761 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25388795 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25388735 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25388780 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25388781 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25388774 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25388759 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25388783 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25388750 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25388680 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25388806 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25388764 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25388723 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25388699 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25388784 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25388730 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25388733 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25388707 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25388704 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25388805 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25388767 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25388757 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25388732 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25388746 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25388755 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25388772 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25388799 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25388765 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25388792 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25388727 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25388721 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25388748 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25388804 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25388802 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25388734 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25388684 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25388719 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25388770 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25388778 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25388787 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25388730 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25388785 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25388769 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25388792 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25388761 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25388783 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25388725 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25388735 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25388767 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06081052 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25388754 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06439747 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25388727 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25388720 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06126845 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25388751 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25388766 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06563680 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25388770 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25388765 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25388738 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25388764 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25388746 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25388781 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25388734 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25388774 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25388757 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25388721 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25388750 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25388755 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25388763 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25388749 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25388772 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05936982 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25388743 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25388780 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06439751 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25388719 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25388790 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25388739 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25388759 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25388794 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25388741 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25388789 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25388778 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25388775 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10566378 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10566377 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 715303 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 360114 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0000360114 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2729333 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2729333 | - | - |
mouse-6 v1.1 (Illumina) | scl30900.1.175_144-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 360114 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl30900.1.175_144-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 360114 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2729333 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2729333 | - | - |
