Assay Details
| Gene Symbol: Adgrf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033493.2 | NP_001028665.2 | 2 - 3 | 359 | 94 | |
| GenBank mRNA | AK142836.1 | - | 2 - 3 | 360 | 94 | |
| BB662982.1 | - | 2 - 3 | 360 | 94 | ||
| BC141353.1 | - | 2 - 3 | 262 | 9055965 | 94 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK142836.1 transcripts.
Set Membership: |
|

Adgrf2
Genomic Map