Assay Details
| Gene Symbol: Cyct | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009989.3 | NP_034119.1 | 2 - 3 | 168 | 87 | |
| GenBank mRNA | AK005582.1 | - | 2 - 3 | 102 | 87 | |
| AK018833.1 | - | 2 - 3 | 102 | 87 | ||
| AK018851.1 | - | 2 - 3 | 102 | 87 | ||
| BC049608.1 | - | 2 - 3 | 122 | 87 | ||
| BC059728.1 | - | 2 - 3 | 164 | 6741773 | 87 | |
| M20625.1 | - | 2 - 3 | 163 | 87 | ||
| X55771.1 | - | 2 - 3 | 101 | 87 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK005582.1, AK018833.1, AK018851.1, BC059728.1, M20625.1, NM_009989.3, X55771.1 transcripts.
Set Membership: |
|

Cyct
Genomic Map