Assay Details
| Gene Symbol: Tspo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027292.2 | NP_081568.1 | 3 - 4 | 424 | 92 | |
| GenBank mRNA | AK011003.1 | - | 3 - 4 | 425 | 92 | |
| BC104344.1 | - | 2 - 3 | 283 | 40049489 | 92 | |
| BC104345.1 | - | 2 - 3 | 283 | 40049491 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P161929 | AK162634.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P161929 | NM_027292.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P161929 | AK011003.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P161929 | AK162634.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P161929 | NM_027292.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P161929 | AK011003.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 58464 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 842209 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 718781 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 41780 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P156365 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P161929 | AK162634.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P161929 | NM_027292.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P161929 | AK011003.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P161929 | AK162634.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P161929 | NM_027292.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P161929 | AK011003.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31217533 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31217524 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31217530 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31217544 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31217528 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31217536 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31217553 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31217556 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31217532 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31217543 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31217547 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31217555 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31217529 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31217540 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31217534 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31217551 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31217527 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31217549 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31217546 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P156365 | - | - |
| Whole Mouse Genome G4122A | A_51_P161929 | AK162634.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P161929 | NM_027292.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P161929 | AK011003.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P161929 | AK162634.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P161929 | NM_027292.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P161929 | AK011003.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P108593 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P156365 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13500531 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13256815 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13135955 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13500533 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13192316 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13256808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217568 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13256815 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217505 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217534 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13500533 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217545 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13398697 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217511 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217528 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217544 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217523 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217547 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217541 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217543 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217551 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13457193 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217532 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13457191 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13500532 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13061425 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217527 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217521 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217529 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13371304 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217530 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217514 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13256813 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217517 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13075901 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10451674 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451673 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451672 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451671 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1453047_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10451670 | - | - |
| Mouse Genome 430 2.0 Array | 1453047_at | - | - |
| Mouse Genome 430 2.0 Array | 1453047_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70026_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70026_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700021061.mm.1 | NM_027292.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700021061.mm.1 | NM_027292.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700021061.mm.1 | TC1700002227.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700021063.mm.1 | NM_027292.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700021063.mm.1 | NM_027292.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700021063.mm.1 | TC1700002227.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700021064.mm.1 | NM_027292.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700021064.mm.1 | NM_027292.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700021064.mm.1 | TC1700002227.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1453047_PM_at | - | - |
| Mouse Expression Set 430 | 1453047_at | - | - |
| Murine Genome U74v2 Set | 103512_PM_at | - | - |
| Murine Genome U74v2 Set | 103512_at | - | - |
| Mouse Genome 430A 2.0 Array | 1453047_at | - | - |
| Mouse Genome 430A 2.0 Array | 1453047_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1453047_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1453047_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 368394 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4480113 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4480113 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004480113 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2635718 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2635718 | - | - |
| mouse-6 v1.1 (Illumina) | scl49830.2.1_22-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4480113 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl49830.2.1_22-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4480113 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2635718 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2635718 | - | - |

Tspo2
Genomic Map