Assay Details
| Gene Symbol: Galk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291002.1 | NP_001277931.1 | 6 - 7 | 709 | 69 | |
| NM_175154.3 | NP_780363.1 | 6 - 7 | 815 | 69 | ||
| GenBank mRNA | AK053002.1 | - | 6 - 7 | 665 | 69 | |
| AK082521.1 | - | 6 - 7 | 706 | 69 | ||
| AK152851.1 | - | 4 - 5 | 398 | 69 | ||
| AK154762.1 | - | 6 - 7 | 664 | 69 | ||
| AK187716.1 | - | 3 - 4 | 316 | 69 | ||
| AK194301.1 | - | 1 - 2 | 39 | 69 | ||
| BC079843.1 | - | 6 - 7 | 679 | 5690056 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Galk2
Genomic Map