Assay Details
| Gene Symbol: Galk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291002.1 | NP_001277931.1 | 7 - 8 | 867 | 76 | |
| NM_175154.3 | NP_780363.1 | 7 - 8 | 973 | 76 | ||
| GenBank mRNA | AK053002.1 | - | 7 - 8 | 823 | 76 | |
| AK082521.1 | - | 7 - 8 | 864 | 76 | ||
| AK152851.1 | - | 5 - 6 | 556 | 76 | ||
| AK154762.1 | - | 7 - 8 | 822 | 76 | ||
| BC079843.1 | - | 7 - 8 | 837 | 5690056 | 76 | |
More Information
Set Membership: |
|

Galk2
Genomic Map