Assay Details
| Gene Symbol: Zswim2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027964.2 | NP_082240.1 | 1 - 2 | 212 | 59 | |
| GenBank mRNA | AK006367.1 | - | 1 - 2 | 213 | 59 | |
| BC120837.1 | - | 1 - 2 | 192 | 40129760 | 59 | |
| BC144860.1 | - | 1 - 2 | 192 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zswim2
Genomic Map