Assay Details
| Gene Symbol: Zswim2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027964.2 | NP_082240.1 | 5 - 6 | 789 | 82 | |
| GenBank mRNA | AK006367.1 | - | 5 - 6 | 790 | 82 | |
| AK017097.1 | - | 5 - 6 | 790 | 82 | ||
| BC120837.1 | - | 5 - 6 | 769 | 40129760 | 82 | |
| BC144860.1 | - | 5 - 6 | 769 | 82 | ||
| EU233996.1 | - | 3 - 4 | 904 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zswim2
Genomic Map