Assay Details
| Gene Symbol: Vegfa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025250.3 | NP_001020421.2 | 2 - 3 | 1138 | 62 | |
| NM_001025257.3 | NP_001020428.2 | 2 - 3 | 1138 | 62 | ||
| NM_001287056.1 | NP_001273985.1 | 2 - 3 | 1138 | 62 | ||
| NM_001287057.1 | NP_001273986.1 | 2 - 3 | 1138 | 62 | ||
| NM_001287058.1 | NP_001273987.1 | 2 - 3 | 1138 | 62 | ||
| NM_009505.4 | NP_033531.3 | 2 - 3 | 1138 | 62 | ||
| GenBank mRNA | AY263146.1 | - | 2 - 3 | 117 | 62 | |
| AY707864.1 | - | 3 - 4 | 335 | 62 | ||
| AY750956.1 | - | 3 - 4 | 296 | 62 | ||
| AY750957.1 | - | 3 - 4 | 259 | 62 | ||
| BC061468.1 | - | 2 - 3 | 511 | 6816435 | 62 | |
| CA321456.1 | - | 2 - 3 | 511 | 62 | ||
| EF028705.1 | - | 3 - 4 | 331 | 62 | ||
| U50279.1 | - | 2 - 3 | 117 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Vegfa
Genomic Map