| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1700020299.mm.1 |
NM_001110266.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020299.mm.1 |
NM_001110266.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020299.mm.1 |
NM_001110267.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020299.mm.1 |
NM_001110267.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020299.mm.1 |
NM_001110268.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020299.mm.1 |
NM_001110268.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020299.mm.1 |
TC1700002162.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700020298.mm.1 |
NM_001110266.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020298.mm.1 |
NM_001110266.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020298.mm.1 |
NM_001110267.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020298.mm.1 |
NM_001110267.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020298.mm.1 |
NM_001110268.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020298.mm.1 |
NM_001110268.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020298.mm.1 |
TC1700002162.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700020301.mm.1 |
NM_001110266.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020301.mm.1 |
NM_001110266.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020301.mm.1 |
NM_001110267.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020301.mm.1 |
NM_001110267.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020301.mm.1 |
NM_001110268.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020301.mm.1 |
NM_001110268.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020301.mm.1 |
TC1700002162.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700020296.mm.1 |
NM_001025250.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020296.mm.1 |
NM_001025250.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020296.mm.1 |
NM_001025257.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020296.mm.1 |
NM_001025257.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020296.mm.1 |
NM_001110266.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020296.mm.1 |
NM_001110266.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020296.mm.1 |
NM_001110267.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020296.mm.1 |
NM_001110267.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020296.mm.1 |
NM_001110268.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020296.mm.1 |
NM_001110268.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020296.mm.1 |
NM_001287056.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020296.mm.1 |
NM_001287057.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020296.mm.1 |
NM_001287058.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020296.mm.1 |
NM_009505.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020296.mm.1 |
NM_009505.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020296.mm.1 |
TC1700002162.mm.1 |
MTA TC |