Assay Details
| Gene Symbol: Mad2l1bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025649.3 | NP_079925.2 | 1 - 2 | 110 | 70 | |
| GenBank mRNA | AK002377.1 | - | 1 - 2 | 96 | 70 | |
| AK004224.1 | - | 1 - 2 | 64 | 70 | ||
| AK156569.1 | - | 1 - 2 | 113 | 70 | ||
| AK220322.1 | - | 1 - 2 | 79 | 70 | ||
| BC005768.1 | - | 1 - 2 | 68 | 2649017 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mad2l1bp
Genomic Map