Assay Details
| Gene Symbol: Cul7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025611.5 | NP_079887.3 | 23 - 24 | 4805 | 94 | |
| GenBank mRNA | AK010907.1 | - | 2 - 3 | 330 | 94 | |
| AK147156.1 | - | 23 - 24 | 4563 | 94 | ||
| AK147905.1 | - | 23 - 24 | 4525 | 94 | ||
| AK172882.1 | - | 22 - 23 | 3878 | 94 | ||
| BC019645.1 | - | 13 - 14 | 2093 | 94 | ||
| BC024345.1 | - | 2 - 3 | 597 | 94 | ||
| BC026946.1 | - | 5 - 6 | 913 | 94 | ||
| BC065096.1 | - | 22 - 23 | 4292 | 94 | ||
| BC100544.1 | - | 2 - 3 | 441 | 94 | ||
| BC138998.1 | - | 23 - 24 | 4743 | 8862020 | 94 | |
| BC139000.1 | - | 23 - 24 | 4743 | 8862022 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cul7
Genomic Map