Assay Details
| Gene Symbol: Polh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030715.3 | NP_109640.1 | 4 - 5 | 726 | 77 | |
| GenBank mRNA | AB027128.1 | - | 4 - 5 | 752 | 77 | |
| AK036296.1 | - | 4 - 5 | 727 | 77 | ||
| BC049159.1 | - | 4 - 5 | 697 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Polh
Genomic Map