Assay Details
| Gene Symbol: Dlk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286013.1 | NP_001272942.1 | 2 - 3 | 288 | 78 | |
| NM_001286028.1 | NP_001272957.1 | 2 - 3 | 175 | 78 | ||
| GenBank mRNA | BC118057.1 | - | 2 - 3 | 175 | 40105170 | 78 |
| BC122518.1 | - | 3 - 4 | 203 | 40105175 | 78 | |
| BY271273.1 | - | 2 - 3 | 293 | 78 | ||
| FM180474.1 | - | 2 - 3 | 288 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dlk2
Genomic Map