Assay Details
| Gene Symbol: Dlk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286013.1 | NP_001272942.1 | 3 - 4 | 346 | 103 | |
| NM_001286028.1 | NP_001272957.1 | 3 - 4 | 233 | 103 | ||
| GenBank mRNA | BC118057.1 | - | 3 - 4 | 233 | 40105170 | 103 |
| BC122518.1 | - | 4 - 5 | 261 | 40105175 | 103 | |
| BG868693.1 | - | 3 - 4 | 326 | 102 | ||
| BY271273.1 | - | 3 - 4 | 351 | 104 | ||
| FM180474.1 | - | 3 - 4 | 346 | 103 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dlk2
Genomic Map