Assay Details
| Gene Symbol: Dnph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207161.3 | NP_997044.2 | 1 - 2 | 182 | 73 | |
| GenBank mRNA | AK146723.1 | - | 1 - 2 | 183 | 73 | |
| AK151589.1 | - | 1 - 2 | 166 | 73 | ||
| BC048355.1 | - | 1 - 2 | 171 | 5388297 | 73 | |
| BC082791.1 | - | 1 - 2 | 208 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dnph1
Genomic Map