Assay Details
| Gene Symbol: Mrps5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029963.2 | NP_084239.1 | 10 - 11 | 938 | 78 | |
| GenBank mRNA | AB049941.1 | - | 10 - 11 | 938 | 78 | |
| AK078198.1 | - | 9 - 10 | 2197 | 78 | ||
| AK083366.1 | - | 10 - 11 | 940 | 78 | ||
| AK155101.1 | - | 10 - 11 | 954 | 78 | ||
| BC021947.1 | - | 10 - 11 | 936 | 5098933 | 78 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mrps5
Genomic Map