Assay Details
| Gene Symbol: Mrps5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029963.2 | NP_084239.1 | 9 - 10 | 873 | 97 | |
| GenBank mRNA | AB049941.1 | - | 9 - 10 | 873 | 97 | |
| AK078198.1 | - | 8 - 9 | 2132 | 97 | ||
| AK083366.1 | - | 9 - 10 | 875 | 97 | ||
| AK155101.1 | - | 9 - 10 | 889 | 97 | ||
| BC021947.1 | - | 9 - 10 | 871 | 5098933 | 97 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mrps5
Genomic Map