Assay Details
| Gene Symbol: Epc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172663.4 | NP_766251.3 | 3 - 4 | 783 | 121 | |
| GenBank mRNA | AK031025.1 | - | 3 - 4 | 756 | 121 | |
| AK047947.1 | - | 3 - 4 | 451 | 121 | ||
| AK141073.1 | - | 74 | 121 | |||
| BC048785.1 | - | 2 - 3 | 222 | 121 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Epc2
Genomic Map