Assay Details
Gene Symbol: Btnl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111094.1 | NP_001104564.1 | 1 - 2 | 263 | 60 | |
GenBank mRNA | BC052925.1 | - | 1 - 2 | 241 | 6398253 | 60 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC052925.1, NM_001111094.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
