Assay Details
Gene Symbol: H2-Eb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010382.2 | NP_034512.2 | 2 - 3 | 452 | 121 | |
GenBank mRNA | AK160485.1 | - | 2 - 3 | 397 | 121 | |
K01145.1 | - | 1 - 2 | 121 | |||
M28408.1 | - | 2 - 3 | 287 | 121 | ||
M28591.1 | - | 1 - 2 | 274 | 121 | ||
M36939.1 | - | 2 - 3 | 121 | |||
M36940.1 | - | 2 - 3 | 372 | 121 | ||
U28490.1 | - | 2 - 3 | 121 | |||
X00958.1 | - | 2 - 3 | 297 | 121 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
