Assay Details
| Gene Symbol: H2-Eb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010382.2 | NP_034512.2 | 3 - 4 | 734 | 98 | |
| GenBank mRNA | AK160485.1 | - | 3 - 4 | 679 | 98 | |
| D90032.1 | - | 3 - 4 | 98 | |||
| M12826.1 | - | 2 - 3 | 98 | |||
| M28591.1 | - | 2 - 3 | 556 | 98 | ||
| M36940.1 | - | 3 - 4 | 654 | 98 | ||
| X00958.1 | - | 3 - 4 | 579 | 98 | ||
| X52642.1 | - | 3 - 4 | 689 | 98 | ||
More Information
Updated Mapping Information
Set Membership: |
|

H2-Eb1
Genomic Map