Assay Details
| Gene Symbol: H2-Eb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010382.2 | NP_034512.2 | 4 - 5 | 851 | 91 | |
| GenBank mRNA | AK155968.1 | - | 4 - 5 | 823 | 91 | |
| AK160485.1 | - | 4 - 5 | 796 | 91 | ||
| BC132163.1 | - | 4 - 5 | 801 | 40130440 | 91 | |
| BC132165.1 | - | 4 - 5 | 804 | 40130442 | 91 | |
| K01145.1 | - | 3 - 4 | 684 | 91 | ||
| M11355.1 | - | 3 - 4 | 686 | 91 | ||
| M12826.1 | - | 3 - 4 | 687 | 91 | ||
| M36939.1 | - | 4 - 5 | 768 | 91 | ||
| M36940.1 | - | 4 - 5 | 771 | 91 | ||
| M73959.1 | - | 1167 | 91 | |||
| M73960.1 | - | 2432 | 91 | |||
| X00958.1 | - | 4 - 5 | 696 | 91 | ||
| X52641.1 | - | 4 - 5 | 776 | 91 | ||
| X52642.1 | - | 4 - 5 | 806 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

H2-Eb1
Genomic Map