Assay Details
| Gene Symbol: Nid2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008695.2 | NP_032721.2 | 20 - 21 | 4283 | 117 | |
| GenBank mRNA | AB017202.1 | - | 21 | 4309 | 117 | |
| AK051664.1 | - | 20 - 21 | 4263 | 117 | ||
| AK083629.1 | - | 20 - 21 | 4260 | 117 | ||
| AK128978.1 | - | 18 - 19 | 3504 | 117 | ||
| AK140832.1 | - | 20 - 21 | 4257 | 117 | ||
| AK164258.1 | - | 20 - 21 | 4254 | 117 | ||
| AY074820.1 | - | 20 - 21 | 4244 | 117 | ||
| BC054746.1 | - | 20 - 21 | 4265 | 6405073 | 117 | |
| BC057016.1 | - | 20 - 21 | 4211 | 5719452 | 117 | |
More Information
Set Membership: |
|

Nid2
Genomic Map