Assay Details
| Gene Symbol: Hmgb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293623.1 | NP_001280552.1 | 3 - 4 | 621 | 75 | |
| NM_001293624.1 | NP_001280553.1 | 3 - 4 | 392 | 75 | ||
| NM_001293625.1 | NP_001280554.1 | 3 - 4 | 388 | 75 | ||
| NM_008253.4 | NP_032279.1 | 3 - 4 | 574 | 75 | ||
| GenBank mRNA | AF022465.1 | - | 2 - 3 | 322 | 75 | |
| AK032163.1 | - | 3 - 4 | 498 | 75 | ||
| AK141399.1 | - | 3 - 4 | 391 | 75 | ||
| AK146795.1 | - | 3 - 4 | 391 | 75 | ||
| AK159119.1 | - | 3 - 4 | 365 | 75 | ||
| AK159322.1 | - | 3 - 4 | 365 | 75 | ||
| BC011276.1 | - | 3 - 4 | 355 | 3980194 | 75 | |
| BC083352.1 | - | 2 - 3 | 306 | 30540227 | 75 | |
| BC158037.1 | - | 3 - 4 | 326 | 9007340 | 75 | |
| CX240626.1 | - | 3 - 4 | 357 | 75 | ||
More Information
Set Membership: |
|

Hmgb3
Genomic Map