Assay Details
| Gene Symbol: Car2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009801.4 | NP_033931.4 | 1 - 2 | 356 | 73 | |
| GenBank mRNA | AK002333.1 | - | 1 - 2 | 91 | 73 | |
| AK146015.1 | - | 1 - 2 | 92 | 73 | ||
| BC044806.1 | - | 1 - 2 | 76 | 73 | ||
| BC055291.1 | - | 1 - 2 | 76 | 6479187 | 73 | |
| BU946040.1 | - | 1 - 2 | 358 | 73 | ||
| K00811.1 | - | 1 - 2 | 57 | 73 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK002333.1, BC055291.1, K00811.1, NM_009801.4 transcripts.
Set Membership: |
|

Car2
Genomic Map