Assay Details
| Gene Symbol: Car2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009801.4 | NP_033931.4 | 3 - 4 | 674 | 75 | |
| GenBank mRNA | AK002333.1 | - | 3 - 4 | 409 | 75 | |
| AK002498.1 | - | 2 - 3 | 143 | 75 | ||
| AK146015.1 | - | 3 - 4 | 410 | 75 | ||
| BC055291.1 | - | 3 - 4 | 394 | 6479187 | 75 | |
| K00811.1 | - | 3 - 4 | 375 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Car2
Genomic Map