Assay Details
| Gene Symbol: Sgpp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030750.3 | NP_109675.1 | 1 - 2 | 817 | 65 | |
| GenBank mRNA | AF247177.1 | - | 1 - 2 | 767 | 65 | |
| AF415215.1 | - | 1 - 2 | 651 | 65 | ||
| AK043586.1 | - | 1 - 2 | 785 | 65 | ||
| AK149860.1 | - | 1 - 2 | 818 | 65 | ||
| AK154116.1 | - | 2 - 3 | 459 | 65 | ||
| BC026814.1 | - | 1 - 2 | 753 | 65 | ||
| BC037592.1 | - | 1 - 2 | 770 | 5004335 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sgpp1
Genomic Map