Assay Details
| Gene Symbol: Ddr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022563.2 | NP_072075.2 | 11 - 12 | 1678 | 67 | |
| GenBank mRNA | AK132504.1 | - | 8 - 9 | 1177 | 67 | |
| BC138826.1 | - | 12 - 13 | 1924 | 8861848 | 67 | |
| BC138827.1 | - | 12 - 13 | 1819 | 8861849 | 67 | |
| G49238.1 | - | 12 - 13 | 1985 | 67 | ||
| X76505.1 | - | 12 - 13 | 1985 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ddr2
Genomic Map