Assay Details
| Gene Symbol: Ddr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022563.2 | NP_072075.2 | 4 - 5 | 592 | 87 | |
| GenBank mRNA | BC138826.1 | - | 5 - 6 | 838 | 8861848 | 87 |
| BC138827.1 | - | 5 - 6 | 733 | 8861849 | 87 | |
| G49238.1 | - | 5 - 6 | 899 | 87 | ||
| X76505.1 | - | 5 - 6 | 899 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ddr2
Genomic Map