Assay Details
| Gene Symbol: Srpx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016911.4 | NP_058607.1 | 1 - 2 | 218 | 125 | |
| GenBank mRNA | AB028049.1 | - | 1 - 2 | 145 | 125 | |
| AK034696.1 | - | 1 - 2 | 220 | 125 | ||
| BC022572.1 | - | 1 - 2 | 174 | 3993025 | 125 | |
| BC027630.1 | - | 1 - 2 | 233 | 125 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Srpx
Genomic Map