Assay Details
| Gene Symbol: Srpx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016911.4 | NP_058607.1 | 3 - 4 | 468 | 65 | |
| GenBank mRNA | AB028049.1 | - | 3 - 4 | 395 | 65 | |
| AK034696.1 | - | 3 - 4 | 470 | 65 | ||
| AK159662.1 | - | 2 - 3 | 457 | 65 | ||
| BC022572.1 | - | 3 - 4 | 424 | 3993025 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Srpx
Genomic Map