Assay Details
Gene Symbol: Brinp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207583.2 | NP_997466.2 | 2 - 3 | 1085 | 98 | |
GenBank mRNA | AK032297.1 | - | 2 - 3 | 1089 | 98 | |
AK045801.1 | - | 2 - 3 | 612 | 98 | ||
AK122549.1 | - | 2 - 3 | 1151 | 98 | ||
BC067038.1 | - | 3 - 4 | 731 | 98 | ||
BC076572.1 | - | 2 - 3 | 1085 | 30363878 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
