Assay Details
| Gene Symbol: Ap3b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009680.3 | NP_033810.2 | 18 - 19 | 2277 | 61 | |
| GenBank mRNA | AF103809.2 | - | 18 - 19 | 2214 | 61 | |
| AK143559.1 | - | 18 - 19 | 2273 | 61 | ||
| AK163996.1 | - | 3 - 4 | 335 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ap3b1
Genomic Map